Order Entry
Canada
ContactUsLinkComponent
Glass Microscope Slide, Frosted
Glass Microscope Slide, Frosted
Catalog # 77663-326
New Product
Glass Microscope Slide, Frosted
Catalog # 77663-326
Supplier Number:  503506
New Product

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    Glass microscope slide, frosted and red ended
  • L×W:
    75×25 mm
  • Thickness:
    1.0 - 1.2 mm
  • Cat. no.:
    77663-326

Specifications

About this item

These clean glass microscope slides with 90° grounded edges. The frosted end slides feature a fine 20 mm frosted area on both sides of one end for easy marking. The red frosted slides feature a 20 mm colored end useful for identifying hazardous materials.

Packaging: Box of 144.