Specifications
- Pack type:Vial
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Mouse
- Size:EA
- Storage conditions:At or below –20 °C
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:N
- Protein/peptide name:Mouse monocyte/DC PB
- Endotoxin level:Low
- Nuclease-free:N
- Grade:RUO
- Shipping temperature:RT
- Cat. no.:77661-204
Specifications
About this item
Cytokines for the differentation of mouse monycyte-derived dendritic cells. Dendritic cells are an important type of immune cell that function to activate T and B cells via their role as an antigen presenting cells. Monocytes isolated from bone marrow can be differentiated to immature dendritic cells (iDCs) through culturing in the presence of interleukin 4 (IL-4) and granulocyte-macrophage colony stimulating factor (GM-CSF). The characteristics of monocyte-derived iDCs are detectable after culture with IL-4/GM-CSF and include: the down regulation of CD14 surface expression, an increase in dextran uptake and an increased response to MIP-1α. Subsequent culturing with pro-inflammatory cytokines (such as TNF-α, IL-1β, and IL-6) can further differentiate the iDCs into mature dendritic cells (mDCs) that have full T cell stimulating capacity. This bundle includes 1x 100 μg of Recombinant Mouse IL-4 and 1x 100 μg of Recombinant Mouse GM-CSF.
- High Quality
- Low Endotoxin
- Affordable
This bundle includes 1×100 μg of Recombinant Mouse IL-4 and 1×100 μg of Recombinant Mouse GM-CSF. Specific concentrations of each chemokine are determined by the end user based on their application and culture conditions.