Order Entry
Canada
ContactUsLinkComponent
 
Human Recombinant TGF-A (Animal free) (from E. coli)
New Product
 
Human Recombinant TGF-A (Animal free) (from E. coli)
New Product

 

  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokine
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    −20 °C
  • Endotoxin content:
    Endotoxin LAL (EU/µg) ≤0.1
  • Biological activity:
    Bioactivity assay: BALB/c 3T3 proliferation, ED50 ≤2 ng/ml, bioactivity ≥5×10⁵ units/mg
  • Gene ID:
    P01135
  • Reconstitution instructions:
    Sterile water at 0.1 mg/ml
  • Endotoxin-free:
    N
  • Carrier-free:
    Y
  • Protease-free:
    N
  • Animal-free:
    Y
  • Protein synonyms:
    TGF-alpha|TGFA|EGF-like TGF, ETGF|TGF type 1
  • Protein/peptide name:
    TGF-A (AF)
  • Purity:
    ≥95%
  • Molecular weight:
    5.6 kDa
  • Sequence:
    VVSHFNDCPD SHTQFCFHGT CRFLVQEDKP ACVCHSGYVG ARCEHADLLA
  • Endotoxin level:
    Low
  • Formulation:
    0.1% Trifluoroacetic Acid (TFA)
  • Nuclease-free:
    N
  • Grade:
    RUO
  • Shipping temperature:
    RT
  • Tested applications:
    Bioassay

 

Frequently Bought Together