Orders LinkContactUsLinkComponent
Sterilization Mesh Cassettes
Sterilization Mesh Cassettes
Sterilization Mesh Cassettes
Certificates
Sterilization Mesh Cassettes
3 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Mesh sterilization baskets are perfect for sterilizing, storing and organizing small components or instrument. These baskets are manufactured from 304 stainlesss steel. They are non-magnetic and electropolished.
  • These baskets come with a wire locking clip
  • This style of sterilization baskets is available in three sizes.
They are burr-free to protect the instrument wraps from tearing. The fine mesh has open squares of 1.6 mm, and the wire edge diameter is 0.7 mm.
Product Family Options
Product Information
  • Dimensions
    Availability
    Price
  • 4×4×2 cm (1.5×1.5×.75")
    Log in for availability
    Log in for availability
    Each
    Each - $90.03
    $90.03
    Each
       $90.03
    Supplier Number: 504766
  • 8×8×3.4 cm (3.1×3.1×1.4")
    Log in for availability
    Log in for availability
    Each
    Each - $99.78
    $99.78
    Each
       $99.78
    Supplier Number: 504767
  • 10.5×7×2.5 cm (4.1×2.75×1")
    Log in for availability
    Log in for availability
    Each
    Each - $138.69
    $138.69
    Each
       $138.69
    Supplier Number: 504768
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.