ContactUsLinkComponent
Surgical Instrument Lubricant
Surgical Instrument Lubricant
Catalog # 77662-132
New Product
Surgical Instrument Lubricant
Catalog # 77662-132
Supplier Number:  500126
New Product
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Surgical instrument lubricant, 240 ml
  • Cat. no.:
    77662-132
Specifications
About this item
The lubricant should be applied after cleaning and before sterilizing instruments. This is a non-silicone based surgical instrument lubricant, which is steam permeable, non-greasy and anti-corrosive.
  • Anticorrosive
  • Steam compatible
  • non-greasy
  • Non-toxic
It prevent spotting, rusting or staining.