Orders LinkContactUsLinkComponent
Mathieu Fine Needle Holders
Mathieu Fine Needle Holders
Mathieu Fine Needle Holders
Certificates
Mathieu Fine Needle Holders
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Mathieu fine needle holders are a precision surgical instrument for microsurgery. They were designed for use in dentistry and oral surgery.
  • Fine, slender, serrated jaws
  • Ergonomic handles
  • Ratchet style lock
  • Tungsten carbide jaws
Ring handles are replaced with ergonomic, spring-style handles. The ratchet lock allows the operator to have a light touch with a secure hold for manipulating a suture needle without having to apply constant pressure on the handles. The design of the locking mechanism reduces hand fatigue. Conventional needle holders have ring handles or spring handles.
Instead of finger rings or traditional spring handles, Mathieu Needle Holders have curved and elongated handles that are designed to be held between the thumb and ring finger. This design provides a more ergonomic grip and better control over the instrument. Mathieu needle holders also have a delicate and slender tip with fine serrations the ensure a firm grip on the suture needle.
Mathieu Needle holders are lighter and slenderer than needle holders like the Mayo-Hegar or Olsen-Hegar needle holders. They are suitable for delicate microsurgical procedures where fine and precise needle control is crucial. Our Mathieu Fine Needle Holders are 14 cm (5.5") long and have tungsten carbide inserts, which improve the usable life of the surgical instrument.
They are manufactured from surgical grade stainless steel or premium German steel, both of which are suitable for autoclaving or chemical sterilization. Always clean your surgical instruments promptly after you use them. Proper cleaning lengthens the usable lifespan of surgical instruments.
Product Family Options
Product Information
  • Length
    Availability
    Price
  • 77662-200
    Mathieu fine needle holder, standard
    Mathieu fine needle holder, standard
    14 cm
    Log in for availability
    Log in for availability
    Each
    Each - $386.86
    $386.86
    Each
       $386.86
    Supplier Number: 500225
  • 77662-202
    Mathieu fine needle holder, german
    Mathieu fine needle holder, german
    14 cm
    Log in for availability
    Log in for availability
    Each
    Each - $306.52
    $306.52
    Each
       $306.52
    Supplier Number: 500225-G
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)