Order Entry
Canada
ContactUsLinkComponent
Laceration Kit
Catalogue # 77663-548
New Product
Laceration Kit
Catalogue # 77663-548
Supplier Number:  504195
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

Spécifications

  • Description:
    Laceration kit
  • Cat. no.:
    77663-548
  • No. of Parts:
    5

Spécifications

À propos de cet article

This 5-piece laceration tool kit contains everything you need for laceration applications.

Delivery: Kit includes:
77661-728 - Needle holder, straight, 14cm, serrated jaws (1)
77662-392 - Forceps, dressing, 12.5cm straight, serrated (1)
77661-948 - Mosquito forceps, 12.5cm straight (2)
77662-800 - Operating scissors, 4.5" straight, sharp/blunt (1)