Entomology Dissection Kit
Catalog # 77663-516
Supplier: WORLD PRECISION INSTRUMENTS LLC
New Product
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Description:Entomology dissection kit
- Cat. no.:77663-516
- No. of Parts:15 pieces
Specifications
About this item
Basic kit for entomology studies.
- Contains 15 pieces
Delivery: Kit includes: 2 teaser needles, straight, 2 teaser needles, angled, 1 iris scissors, 11.5 cm, 1 scalpel handle #3, 1 adson micro forceps, 12 cm, 1 dropper, 1 micro dissection forceps, 12 cm, straight, 1 micro dissection forceps, 12 cm, curved, 1 entomology forceps, 10 cm, 1 tweezers with magnifying mirror, 1 stainless steel ruler, 6", 10 blades #10, 1 instrument storage portfolio. Case included.