LED Ring Light
Product Details & Documents
'White' light illumination with 72 LED bulbs.
- Maximum opening 61 mm
- Brightness adjustable
- ESD safe
- Direct top illumination plus four zones for shadows
- Low cost
- Less bulky than halogen
- Cold light with no heat
Ring light is divided into four areas and each area is turned on and off separately.
Application: Stereo microscope use
Specifications
- Description:LED ring light, US plug
- Electrical:90 - 264 VAC, 50/60 Hz
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)