Orders LinkContactUsLinkComponent
Weitlaner Retractors, Self-Retaining
Weitlaner Retractors, Self-Retaining
Weitlaner Retractors, Self-Retaining
Weitlaner Retractors, Self-Retaining
A weitlaner retractor is a self-retaining, handheld surgical instrument used to hold soft tissues apart during surgical procedures to give the surgeon access to the site. The retractor is self-retaining, meaning it remains in place once it is positioned, freeing your hands.
  • 10.2 cm (4") Long
  • 2×3 Blunt or sharp prongs
  • 3 or 4 cm Maximum spread
  • Ratchet locking mechanism for consistent hold and hands-free operation
  • Ring handle for comfortable grip
  • Surgical grade stainless steel or German steel
The weitlaner retractor has two parallel blades with prongs, each with blunt or sharp, curved, and slightly tapered ends. The retractor has a ratchet locking mechanism to secure the blades with the desired spread. This mechanism enables easy adjustment of the retraction angle and provides stability during the procedure. The ring handles give you a firm, comfortable grip.
These weitlaner retractors with two prongs on one blade and three on the other. WPI recommends properly cleaning all your surgical instruments promptly after use. This improves the useable lifespan of your surgical tools.
Once positioned and locked in place, a self-retaining retractor holds the tissues or structures in the desired position without requiring continuous attention. This frees your hands for other critical tasks during the procedure. By holding the tissues or organs away from the surgical site, a self-retaining retractor offers you enhanced visibility and greater access to the surgical site.
The ratchet or locking mechanism in a self-retaining retractor ensures a consistent and controlled level of tissue retraction throughout the procedure. This stability helps maintain the exposure required for you to work effectively. Holding and retracting tissues manually for an extended period can lead to hand (and arm) fatigue. With a self-retaining retractor, you can avoid prolonged physical strain, as the retractor holds the tissues in place without continuous effort.
Product Family Options
  • Description
    Length
  • Weitlaner retractor, self-retaining, 3 cm maximum spread, blunt
    10.2 cm (4")
    Each
    Each - $347.93
    $347.93
    Each
       $347.93
     : 501328
  • Weitlaner retractor, self-retaining, 3 cm maximum spread, blunt, german
    10.2 cm (4")
    Each
    Each - $409.20
    $409.20
    Each
       $409.20
     : 501328-G
  • Weitlaner retractor, self-retaining, 4 cm maximum spread, sharp
    10.2 cm (4")
    Each
    Each - $304.15
    $304.15
    Each
       $304.15
     : 501314
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)