About this item
Rubber-tipped forceps for handling rodents are designed for delicate and precise handling of small animals like mice and rats. These forceps feature a unique design with rubber tips to ensure a secure yet gentle grip, reducing the risk of injury or discomfort to the animals.
- Rodent handling forceps
- Replaceable rubber tips are gentle
- Excellent for small animal handling
Rubber-tipped forceps are made from surgical grade stainless steel, which provides durability and strength. The ringed tips of these forceps have replaceable inserts made of soft, non-slip rubber. The rubber grippers ensure a secure grasp without causing damage to the skin or fur of the animals. The forceps are 21 cm (8.25") long and are straight. They are designed to provide precise control over the animal without causing unnecessary stress or harm. The rubber tips offer a non-slip grip, preventing the animal from slipping or being injured during handling. This feature is particularly useful when holding rodents for various procedures, such as injections, examinations, or minor surgeries.
Rubber tipped rodent forceps are sterilizable. The stainless steel components can withstand sterilization processes such as autoclaving, ensuring hygiene and preventing cross-contamination between animals. And replacement rubber tips are also available. These forceps are an essential tool in animal research facilities, veterinary clinics, and laboratory settings where precise and gentle handling of rodents is crucial for various experimental or medical procedures. The rubber tips help maintain a secure grip without causing harm, ensuring the well-being of the animals during experimentation, treatments, or examinations.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.