About this item
The AirTherm™ SMT is a new generation of heat control system from WPI designed to be used in Live Cell Imaging applications with microscopes fitted with a full microscope environmental chamber enclosure. The standard AirTherm™ SMT controls temperature and, as an option, monitors humidity.
- Precision heat controller for use in live cell imaging and custom incubators
- Control heat and monitor humidity (optional) with a single controller
- Electrically and acoustically quiet
- Quick, precise response to thermal change
Benefits:
PID control algorithms allow for tight control of temperature in the environmental chamber
System includes hoses and probes so that it is ready to connect with your microscope chamber, incubator or controlled environment
Applications:
Live cell imaging systems fitted with full microscope environmental chamber enclosures
Control environment in custom incubators
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Catalog No:
- 77664-042
- 77664-040
- Temperature accuracy:
- 0.2 °C
- 0.2 °C
- Sensor:
- Platinum RTD 1000 Ω
- Platinum RTD 1000 Ω