USB Cable, 3M Locking
Product Details & Documents
Replacement USB3 cable.
For use with ¹/₁.₈" color CCD (USBCAM202), ¹/₂" color CCD (USBCAM152), ¹/₃" color CCD (USBCAM133).
Specifications
- Description:Replacement USB3 cable
- Length:3'
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)