ContactUsLinkComponent
LiquiVet Rapid Tissue Adhesive
LiquiVet Rapid Tissue Adhesive
Catalog # 77663-686
New Product
undefined
LiquiVet Rapid Tissue Adhesive
Catalog # 77663-686
Supplier Number:  504561
New Product
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    LiquiVet rapid tissue adhesive
  • Cat. no.:
    77663-686
Specifications
About this item
LiquiVet monomeric formulation forms a thin, waterproof adhesion to bond tissue in a variety of vet procedures. Comes in 3 g dispensing bottles with five applicators.
  • Fast-acting cyanoacrylate-based tissue adhesive for wound care
  • Polymerizes in 2 seconds, ideal for skin closure
  • Superior performance in the time necessary to close a wound
Excellent for quick repair of minor lacerations, abrasions, and other minor wounds.
For external lacerations, not to be used internally.
Caution: Veterinary use only.