Mouse Th9 Cell Polarizing Recombinant Protein Bundle (rm IL-4, rh TGF-β 1) (from E. coli)
Catalog # 77661-228
Supplier: FUJIFILM IRVINE SCIENTIFIC, INC
New Product
CAS Number:
Specifications
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Mouse
- Size:20 μg (IL-4) 5 μg (TGF-β 1)
- Storage conditions:At or below –20 °C
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:N
- Protein/peptide name:Mouse TH9 cell polarizing PB
- Endotoxin level:Low
- Nuclease-free:N
- Grade:RUO
- Shipping temperature:RT
- Cat. no.:77661-228
Specifications
About this item
Cytokines for polarizing mouse T cells into Th9 cells. Th9 cells are a unique subset of T helper cells that produce IL-9. Although the exact function of IL-9 producing T cells is yet to be completely clear it is proposed that these cells could be important in inflammation and helminth infections. Th9 cells can be generated from naive CD4+ T cells in culture by culturing them in the presence of IL-4 and TGF-β 1. It also appears that IFNγ and IL-27 can inhibit the generation of Th9 cells.
- High Quality
- Low Endotoxin
- Affordable
Caution: For research use only.