Order Entry
Canada
ContactUsLinkComponent
Human/Mouse Recombinant IHH (from E. coli)
Human/Mouse Recombinant IHH (from E. coli)
New Product
Certificates
Human/Mouse Recombinant IHH (from E. coli)
New Product

Specifications

  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokine
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
    Mouse
  • Storage conditions:
    At or below –20 °C
  • Endotoxin content:
    Endotoxin LAL (EU/µg) ≤0.1
  • Biological activity:
    Bioactivity: No biological activity data is available at this time
  • Gene ID:
    Q14623 / P97812
  • Amino acid number:
    177
  • Reconstitution instructions:
    10 mM HCl at 0.1 mg/ml
  • Endotoxin-free:
    N
  • Carrier-free:
    Y
  • Protease-free:
    N
  • Animal-free:
    N
  • Protein synonyms:
    HHG-2
  • Protein/peptide name:
    H/M IHH
  • Purity:
    ≥95%
  • Molecular weight:
    19.9 kDa
  • Sequence:
    MIIGPGRVVG SRRRPPRKLV PLAYKQFSPN VPEKTLGASG RYEGKIARSS ERFKELTPNY NPDIIFKDEE NTGADRLMTQRCKDRLNSLA ISVMNQWPGV KLRVTEGWDE DGHHSEESLH YEGRAVDITT SDRDRNKYGL LARLAVEAGF DWVYYESKAH VHCSVKSEHSAAAKTGG
  • Endotoxin level:
    Low
  • Formulation:
    0.1% Trifluoroacetic acid (TFA)
  • Nuclease-free:
    N
  • Grade:
    RUO
  • Shipping temperature:
    RT

Specifications

Frequently Bought Together