Order Entry
Canada
Orders LinkContactUsLinkComponent
Silicone Mat

Silicone Mat

Silicone mat, 38×25.5 cm (15×10")
Catalog # 77662-250
Supplier #:  500255
Silicone Mat
Catalog # 77662-250
Silicone Mat

Silicone mat, 38×25.5 cm (15×10")

Price
$289.54
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

This silicone mat can protect delicate surgical instruments by gently (but firmly) cushioning them and helping to keep them in place to prevent damages.

  • Autoclavable

Delivery information: Instruments sold separately.

Specifications
  • Description:
    Silicone mat, 38×25.5 cm (15×10")

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.