Mouse/Rat Recombinant Rantes/CCL5 (from E. coli)
Fournisseur: FUJIFILM IRVINE SCIENTIFIC, INC
New Product
Certificats
À propos de cet article
Regulated upon activation, normal T cell expressed and secreted (RANTES), also called CCL5, is a chemokine produced by T cells three to five days after T cell activation. RANTES signals through G protein-coupled receptors CCR5, CCR3, CCR1, and through the human CMV-encoded viral receptor US28. RANTES functions to recruit immune cells to inflammatory sites.
- High quality
- Low endotoxin
- Affordable
Caution: For research use only.
Spécifications
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:RatMouse
- Storage conditions:At or below –20 °C
- Endotoxin content:Endotoxin LAL (EU/µg) ≤0.1
- Biological activity:Bioactivity Assay: THP-1 chemotaxis, ED₅₀ ≤250 ng/ml, Bioactivity ≥NA
- Gene ID:Q5XZF2 / P50231
- Reconstitution instructions:Sterile water at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:N
- Protein synonyms:CCL5
- Protein/peptide name:M/R RANTES / CCL5
- Purity:≥95%
- Molecular weight:7.9 kDa
- Sequence:SPYGSDTTPC CFAYLSLALP RAHVKEYFYT SSKCSNLAVV FVTRRNRQVC ANPEKKWVQE YINYLEMS
- Endotoxin level:Low
- Formulation:0.1% Trifluoroacetic acid (TFA)
- Nuclease-free:N
- Grade:RUO
- Shipping temperature:RT
Spécifications
Frequently Bought Together