Order Entry
Canada
ContactUsLinkComponent
 
Human Recombinant rh GM-CSF (GMP) (from E. coli)
New Product
 
Human Recombinant rh GM-CSF (GMP) (from E. coli)
New Product

 

  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokine
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    At or below –20 °C
  • Endotoxin content:
    Endotoxin LAL (EU/µg) ≤0.05
  • Biological activity:
    Bioactivity assay: Proliferation of TF-1, ED50 ≤0.2 ng/ml, Bioactivity ≥1.6×10⁴ IU/µg
  • Gene ID:
    P04141
  • Reconstitution instructions:
    Sterile water at 0.1 - 0.5 mg/ml
  • Endotoxin-free:
    N
  • Carrier-free:
    Y
  • Protease-free:
    N
  • Animal-free:
    Y
  • Protein synonyms:
    MGI-1GM|CSF-2|Pluripoietin-α
  • Protein/peptide name:
    GM-CSF
  • Purity:
    ≥98%
  • Molecular weight:
    14.6 kDa
  • Sequence:
    MAPARSPSPS TQPWEHVNAI QEARRLLNLS RDTAAEMNET VEVISEMFDL QEPTCLQTRL ELYKQGLRGS LTKLKGPLTM MASHYKQHCP PTPETSCATQ IITFESFKEN LKDFLLVIPF DCWEPVQE
  • Endotoxin level:
    Low
  • Formulation:
    10 mM sodium phosphate, pH 7.5
  • Nuclease-free:
    N
  • Grade:
    CTGrade GMP
  • Shipping temperature:
    Ambient
  • Tested applications:
    Bioassay

 

Frequently Bought Together