ContactUsLinkComponent
Anesthesia Facemask for Cunningham Stereotaxic Frame Adapters
Anesthesia Facemask for Cunningham Stereotaxic Frame Adapters
Catalog # 77663-050
New Product
Anesthesia Facemask for Cunningham Stereotaxic Frame Adapters
Catalog # 77663-050
Supplier Number:  502063-SGM
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Anesthesia Facemask, small
  • Environmentally Preferable:
  • Cat. no.:
    77663-050
  • For Use With:
    Cunningham stereotaxic adapter
Specifications
About this item
Our custom toothbar for Cunningham adaptor is designed to accommodate use of a soft gas mask. The included mask securely fits the toothbar to eliminate gas leakage.
  • Facemask custom fitted to a toothbar that fits a cunningham stereotaxic adapter
  • Anatomically correct design for a tight fit for mice and rats
  • Zero leakage
  • Zero dead space
  • Ports swivel 360°
  • Optically clear
  • Designed to provide access to eyes and head of the rodent
  • Mask provides stability of the animal for examinations and surgery
  • Includes OC-tubing and OC-adapter (requires MSPP-EZ-1130)
The OC gas masks allow you access to the eyes and head of the rodent while sealing the nose and mouth. The gas masks provides stability of the animal for examinations and surgery, and they can be easily incorporated in existing examination proedures. The waste-line connects to gravity or vacuum filters, and swiveling ports allow for greater flexibility in positions. The are easily adapted for stereotaxic applications.
The face mask reduces the technicians exposure to gas by sequestering and filtering waste, minimizing dead space and leakage. Can reduce rodent stress by limiting handling required when using injectable anesthesia. By choosing these gas masks with isoflurane and thereby reducing the use of Schedule II drugs, you can simplify your lab management. Setting-up and changing masks is simple, and the swivel adapters allow you to make adjustments quickly and even modify the mask for stereotaxic uses. Gas anesthesia reduces recovery time, making for a faster and more efficient lab.