These Vannas scissors are specialized spring scissors designed for precision cutting and often used by scientific researchers.
- Straight blades
- 0.015×0.015 mm super fine tips
- 3 mm blades
- Stainless steel
- Autoclavable
Compared with other spring scissors, Vannas scissors are distinguished by their delicate, thin, sharp tips. The slender, pointed tips allows for precise cutting with minimal damage to surrounding tissues. Manufactured with surgical grade stainless steel, these scissors are durable and resistant to corrosion. As with all your surgical instruments, these scissors should be cleaned immediately after use to ensure a long service life and minimize staining or corrosion.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.