Orders LinkContactUsLinkComponent
Tissue Forceps
Tissue Forceps
15 cm (6"), Fine
Catalog # 77662-762
Supplier #:  501742-G
Tissue Forceps
Catalog # 77662-762
Tissue Forceps
15 cm (6"), Fine
Price
$64.78
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Tissue forceps with 1×2 teeth have one tooth on one tip and two teeth on the other tip of the tweezers. These tweezers have a fine tip for delicate tissue manipulation, such as during ophthalmic surgery or plastic surgery.
  • Straight
  • Spring tension shank
  • 1×2 Teeth for a fine tip with a firm grasp
  • Premium German stainless steel
Laboratory researchers choose these tissue forceps for precision work and microdissection. The teeth ensure a firm grasp of the tissue, which is ideal for slippery or hard to hold tissues. While tweezers with teeth run the risk of puncturing delicate tissue, the 1×2 configuration causes less tissue trauma than surgical tweezers with more teeth.
These have straight tips and are 15 cm (6") long and fit easily into your hand for precision work. These are manufactured with premium grade German stainless steel, which is suitable for sterilization by autoclave or chemical sterilant. As with all fine surgical instruments, for best results and a long life for the instruments, always clean the surgical forceps thoroughly after each use.
Specifications
  • Length:
    15 cm (6")
  • Tip style:
    Fine
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)