Human Recombinant FGF-9 (Animal free) (from E. coli)
New Product
Fibroblast growth factor 9 (FGF-9) is a mitogen and survival factor for nerve and mesenchymal cells. FGF-9 functions as an autocrine and paracrine factor to support the growth and survival of motor neurons and prostate tissue. FGF-9 expression in the gonad is also necessary for sex determination.
- High Quality
- Low Endotoxin
- Affordable
: For research use only.
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:–20 °C
- Endotoxin content:Endotoxin LAL (EU/µg) ≤0.1
- Biological activity:Bioactivity assay: NR6R 3T3 cell proliferation, ED50 ≤2 ng/ml, Bioactivity ≥5.0×10⁵ units/mg
- Gene ID:P31371
- Amino acid number:207
- Reconstitution instructions:Sterile water at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:Y
- Protein synonyms:Heparin-binding growth factor -9 (HBGF-9)|Glia activating factor (GAF)
- Protein/peptide name:FGF-9
- Purity:≥95%
- Molecular weight:23.4 kDa
- Sequence:MPLGEVGNYF GVQDAVPFGN VPVLPVDSPV LLSDHLGQSE AGGLPRGPAV TDLDHLKGIL RRRQLYCRTG FHLEIFPNGT IQGTRKDHSR FGILEFISIA VGLVSIRGVD SGLYLGMNEK GELYGSEKLT QECVFREQFE ENWYNTYSSN LYKHVDTGRR YYVALNKDGT PREGTRTKRH QKFTHFLPRP VDPDKVPELY KDILSQS
- Endotoxin level:Low
- Formulation:10 mM sodium phosphate, 25 mM sodium chloride, 50 mM sodium sulfate, pH 7.5
- Nuclease-free:N
- Grade:RUO
- Shipping temperature:RT
- Tested applications:Bioassay
Frequently Bought Together