About this item
Vannas Tübingen scissors are specialized microsurgical scissors that are highly precise and delicate, commonly used in ophthalmic and neurological procedures. They are named after Tübingen, a city in Germany, where the Max Planck Institute for Biological Cybernetics is located.
- Ambidextrous spring scissors for delicate surgeries
- Curved or straight blads
- Double spring thumb scissors
- German manufactured
Vannas Tübingen scissors are incredibly fine instruments, crafted of high-quality surgical grade stainless steel with extreme precision. Their design includes thin, and sharp blades, which allow for highly accurate and precise cutting in microsurgical settings. The tips of these scissors are exceptionally sharp and thin, enabling them to access and cut very fine and delicate tissues without causing unnecessary damage to surrounding structures. This is especially crucial in delicate surgeries, such as in ophthalmic procedures, where precision is paramount.
The blades of Vannas Tübingen scissors can be curved or straight. This curvature allows for better access and visibility in tight spaces or when working around delicate structures. These spring scissors are ambidextrous by nature, often preferred by left handed people. Like most surgical instruments, Vannas Tübingen scissors are designed to be sterilizable, allowing for safe and hygienic use in surgical settings.
The combination of their ultra-fine, sharp tips, curved design, and exceptional precision makes Vannas Tübingen scissors essential tools in microsurgical settings, particularly in delicate eye surgeries and neurological procedures where the utmost precision and care are required to ensure successful outcomes with minimal damage to surrounding tissues.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.