Orders LinkContactUsLinkComponent
Micro Castroviejo Needle Holders
Micro Castroviejo Needle Holders
Micro Castroviejo Needle Holders
Certificates
Micro Castroviejo Needle Holders
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Micro Castroviejo Needle Holders are a surgical instrument used for microsurgery requiring precise control and manipulation of small needles, though they are most commonly used in ophthalmic surgery. These have a fine rounded tip. Our Castroviejo Needle Holders feature fine tips for grasping and manipulation of small, delicate needles.
  • 0.5 cm round tips with diamond dust
The jaws of these Micro Castroviejo Needle Holders are diamond dusted for a secure grasp of fine needles. These Catroviejo needle holders have a spring action for situations when you need constant pressure on the needle and precision control. These are useful when working under a microscope or in confined spaces.
They are an excellent choice for left-handed operators, too, as they are designed for ambidextrous use. Be sure to clean, disinfect, and sterilize your surgical instruments promptly after each use to ensure a long life for your surgical tools. These Castroviejo Needle Holders are available in surgical grade stainless steel, and they are autoclavable.
Product Family Options
Product Information
  • Material
    Length
    Curvature
    Availability
    Price
  • Stainless steel
    9 cm
    Curved
    Log in for availability
    Log in for availability
    Each
    Each - $557.17
    $557.17
    Each
       $557.17
    Supplier Number: 503377
  • Stainless steel
    9 cm
    Straight
    Log in for availability
    Log in for availability
    Each
    Each - $557.17
    $557.17
    Each
       $557.17
    Supplier Number: 503376
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.