Mouse Recombinant TNF-A (from E. coli)
About this item
Tumor necrosis factor alpha (TNF-α) is an inflammatory cytokine secreted by macrophages, monocytes, neutrophils, T cells, and NK-cells following stimulation by bacterial lipopolysaccharide (LPS). TNF-α signal activation occurs through two receptors, TNFR1 and TNFR2. TNFR1 is expressed on most cell types, unlike TNFR2, which is expressed mainly on immune cells. TNF-α functions to stimulate phagocytosis in macrophages, chemoattract neutrophils, increase insulin resistance, and induce fever.
- High quality
- Low endotoxin
- Affordable
5 Options Available Below
Product Details & Documents
Tumor necrosis factor alpha (TNF-α) is an inflammatory cytokine secreted by macrophages, monocytes, neutrophils, T cells, and NK-cells following stimulation by bacterial lipopolysaccharide (LPS). TNF-α signal activation occurs through two receptors, TNFR1 and TNFR2. TNFR1 is expressed on most cell types, unlike TNFR2, which is expressed mainly on immune cells. TNF-α functions to stimulate phagocytosis in macrophages, chemoattract neutrophils, increase insulin resistance, and induce fever.
- High quality
- Low endotoxin
- Affordable
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- 77660-840
- 77660-836
- 77660-832
- 77660-838
- 77660-834
- Protein/peptide name:
- TNF-A (AF)
- TNF-A (AF)
- TNF-A (AF)
- TNF-A (AF)
- TNF-A (AF)
- Protein synonyms:
- Cytotoxin|DIF|Necrosin|Cachexin|TNFSF2|TNF|Cachectin
- Cytotoxin|DIF|Necrosin|Cachexin|TNFSF2|TNF|Cachectin
- Cytotoxin|DIF|Necrosin|Cachexin|TNFSF2|TNF|Cachectin
- Cytotoxin|DIF|Necrosin|Cachexin|TNFSF2|TNF|Cachectin
- Cytotoxin|DIF|Necrosin|Cachexin|TNFSF2|TNF|Cachectin
- Protein/peptide type:
- Recombinant
- Recombinant
- Recombinant
- Recombinant
- Recombinant
- Protein Function:
- Cytokine
- Cytokine
- Cytokine
- Cytokine
- Cytokine
- Species:
- Mouse
- Mouse
- Mouse
- Mouse
- Mouse
- Source:
- E. coli
- E. coli
- E. coli
- E. coli
- E. coli
- Conjugation:
- Unconjugated
- Unconjugated
- Unconjugated
- Unconjugated
- Unconjugated
- Gene ID:
- P06804
- P06804
- P06804
- P06804
- P06804
- Sequence:
- MLRSSSQNSS DKPVAHVVAN HQVEEQLEWL SQRANALLAN GMDLKDNQLV VPADGLYLVY SQVLFKGQGC PDYVLLTHTV SRFAISYQEK VNLLSAVKSP CPKDTPEGAE LKPWYEPIYL GGVFQLEKGD QLSAEVNLPK YLDFAESGQV YFGVIAL
- MLRSSSQNSS DKPVAHVVAN HQVEEQLEWL SQRANALLAN GMDLKDNQLV VPADGLYLVY SQVLFKGQGC PDYVLLTHTV SRFAISYQEK VNLLSAVKSP CPKDTPEGAE LKPWYEPIYL GGVFQLEKGD QLSAEVNLPK YLDFAESGQV YFGVIAL
- MLRSSSQNSS DKPVAHVVAN HQVEEQLEWL SQRANALLAN GMDLKDNQLV VPADGLYLVY SQVLFKGQGC PDYVLLTHTV SRFAISYQEK VNLLSAVKSP CPKDTPEGAE LKPWYEPIYL GGVFQLEKGD QLSAEVNLPK YLDFAESGQV YFGVIAL
- MLRSSSQNSS DKPVAHVVAN HQVEEQLEWL SQRANALLAN GMDLKDNQLV VPADGLYLVY SQVLFKGQGC PDYVLLTHTV SRFAISYQEK VNLLSAVKSP CPKDTPEGAE LKPWYEPIYL GGVFQLEKGD QLSAEVNLPK YLDFAESGQV YFGVIAL
- MLRSSSQNSS DKPVAHVVAN HQVEEQLEWL SQRANALLAN GMDLKDNQLV VPADGLYLVY SQVLFKGQGC PDYVLLTHTV SRFAISYQEK VNLLSAVKSP CPKDTPEGAE LKPWYEPIYL GGVFQLEKGD QLSAEVNLPK YLDFAESGQV YFGVIAL
- Amino acid number:
- 157
- 157
- 157
- 157
- 157
- Protease-free:
- N
- N
- N
- N
- N
- Animal-free:
- Y
- Y
- Y
- Y
- Y
- Nuclease-free:
- N
- N
- N
- N
- N
- Carrier-free:
- Y
- Y
- Y
- Y
- Y
- Endotoxin-free:
- N
- N
- N
- N
- N
- Endotoxin level:
- Low
- Low
- Low
- Low
- Low
- Endotoxin content:
- Endotoxin LAL (EU/µg) ≤0.1
- Endotoxin LAL (EU/µg) ≤0.1
- Endotoxin LAL (EU/µg) ≤0.1
- Endotoxin LAL (EU/µg) ≤0.1
- Endotoxin LAL (EU/µg) ≤0.1
- Biological activity:
- Bioactivity Assay: Cytolysis of mouse L929 cells in the presence of Actinomycin D, ED₅₀ ≤0.1 ng/ml, Bioactivity ≥1.0×10⁷ units/mg
- Bioactivity Assay: Cytolysis of mouse L929 cells in the presence of Actinomycin D, ED₅₀ ≤0.1 ng/ml, Bioactivity ≥1.0×10⁷ units/mg
- Bioactivity Assay: Cytolysis of mouse L929 cells in the presence of Actinomycin D, ED₅₀ ≤0.1 ng/ml, Bioactivity ≥1.0×10⁷ units/mg
- Bioactivity Assay: Cytolysis of mouse L929 cells in the presence of Actinomycin D, ED₅₀ ≤0.1 ng/ml, Bioactivity ≥1.0×10⁷ units/mg
- Bioactivity Assay: Cytolysis of mouse L929 cells in the presence of Actinomycin D, ED₅₀ ≤0.1 ng/ml, Bioactivity ≥1.0×10⁷ units/mg
- Purity:
- ≥95%
- ≥95%
- ≥95%
- ≥95%
- ≥95%
- Grade:
- RUO
- RUO
- RUO
- RUO
- RUO
- Formulation:
- 10 mM sodium phosphate, 50 mM sodium chloride, pH 7.5
- 10 mM sodium phosphate, 50 mM sodium chloride, pH 7.5
- 10 mM sodium phosphate, 50 mM sodium chloride, pH 7.5
- 10 mM sodium phosphate, 50 mM sodium chloride, pH 7.5
- 10 mM sodium phosphate, 50 mM sodium chloride, pH 7.5
- Molecular weight:
- 17.4 kDa
- 17.4 kDa
- 17.4 kDa
- 17.4 kDa
- 17.4 kDa
- Reconstitution instructions:
- Sterile water at 0.1 mg/ml
- Sterile water at 0.1 mg/ml
- Sterile water at 0.1 mg/ml
- Sterile water at 0.1 mg/ml
- Sterile water at 0.1 mg/ml
- Storage conditions:
- –20 °C
- –20 °C
- –20 °C
- –20 °C
- –20 °C
- Shipping temperature:
- RT
- RT
- RT
- RT
- RT
- Tested applications:
- Bioassay
- Bioassay
- Bioassay
- Bioassay
- Bioassay
Specifications
Product Family Options
Product Information
- Pack typeSizeAvailabilityPrice
- Supplier Number: 200-31AF-5UGSpecifications:
More
Specifications
Cat. No.77660-840Protein/peptide nameTNF-A (AF)Protein synonymsCytotoxin|DIF|Necrosin|Cachexin|TNFSF2|TNF|CachectinProtein/peptide typeRecombinantProtein FunctionCytokineSpeciesMouseSourceE. coliConjugationUnconjugatedGene IDP06804SequenceMLRSSSQNSS DKPVAHVVAN HQVEEQLEWL SQRANALLAN GMDLKDNQLV VPADGLYLVY SQVLFKGQGC PDYVLLTHTV SRFAISYQEK VNLLSAVKSP CPKDTPEGAE LKPWYEPIYL GGVFQLEKGD QLSAEVNLPK YLDFAESGQV YFGVIALAmino acid number157Protease-freeNAnimal-freeYNuclease-freeNCarrier-freeYEndotoxin-freeNEndotoxin levelLowEndotoxin contentEndotoxin LAL (EU/µg) ≤0.1Biological activityBioactivity Assay: Cytolysis of mouse L929 cells in the presence of Actinomycin D, ED₅₀ ≤0.1 ng/ml, Bioactivity ≥1.0×10⁷ units/mgPurity≥95%GradeRUOFormulation10 mM sodium phosphate, 50 mM sodium chloride, pH 7.5Molecular weight17.4 kDaReconstitution instructionsSterile water at 0.1 mg/mlStorage conditions–20 °CShipping temperatureRTTested applicationsBioassay - Supplier Number: 200-31AF-20UGSpecifications:
More
Specifications
Cat. No.77660-836Protein/peptide nameTNF-A (AF)Protein synonymsCytotoxin|DIF|Necrosin|Cachexin|TNFSF2|TNF|CachectinProtein/peptide typeRecombinantProtein FunctionCytokineSpeciesMouseSourceE. coliConjugationUnconjugatedGene IDP06804SequenceMLRSSSQNSS DKPVAHVVAN HQVEEQLEWL SQRANALLAN GMDLKDNQLV VPADGLYLVY SQVLFKGQGC PDYVLLTHTV SRFAISYQEK VNLLSAVKSP CPKDTPEGAE LKPWYEPIYL GGVFQLEKGD QLSAEVNLPK YLDFAESGQV YFGVIALAmino acid number157Protease-freeNAnimal-freeYNuclease-freeNCarrier-freeYEndotoxin-freeNEndotoxin levelLowEndotoxin contentEndotoxin LAL (EU/µg) ≤0.1Biological activityBioactivity Assay: Cytolysis of mouse L929 cells in the presence of Actinomycin D, ED₅₀ ≤0.1 ng/ml, Bioactivity ≥1.0×10⁷ units/mgPurity≥95%GradeRUOFormulation10 mM sodium phosphate, 50 mM sodium chloride, pH 7.5Molecular weight17.4 kDaReconstitution instructionsSterile water at 0.1 mg/mlStorage conditions–20 °CShipping temperatureRTTested applicationsBioassay - Supplier Number: 200-31AF-100UGSpecifications:
More
Specifications
Cat. No.77660-832Protein/peptide nameTNF-A (AF)Protein synonymsCytotoxin|DIF|Necrosin|Cachexin|TNFSF2|TNF|CachectinProtein/peptide typeRecombinantProtein FunctionCytokineSpeciesMouseSourceE. coliConjugationUnconjugatedGene IDP06804SequenceMLRSSSQNSS DKPVAHVVAN HQVEEQLEWL SQRANALLAN GMDLKDNQLV VPADGLYLVY SQVLFKGQGC PDYVLLTHTV SRFAISYQEK VNLLSAVKSP CPKDTPEGAE LKPWYEPIYL GGVFQLEKGD QLSAEVNLPK YLDFAESGQV YFGVIALAmino acid number157Protease-freeNAnimal-freeYNuclease-freeNCarrier-freeYEndotoxin-freeNEndotoxin levelLowEndotoxin contentEndotoxin LAL (EU/µg) ≤0.1Biological activityBioactivity Assay: Cytolysis of mouse L929 cells in the presence of Actinomycin D, ED₅₀ ≤0.1 ng/ml, Bioactivity ≥1.0×10⁷ units/mgPurity≥95%GradeRUOFormulation10 mM sodium phosphate, 50 mM sodium chloride, pH 7.5Molecular weight17.4 kDaReconstitution instructionsSterile water at 0.1 mg/mlStorage conditions–20 °CShipping temperatureRTTested applicationsBioassay - Supplier Number: 200-31AF-500UGSpecifications:
More
Specifications
Cat. No.77660-838Protein/peptide nameTNF-A (AF)Protein synonymsCytotoxin|DIF|Necrosin|Cachexin|TNFSF2|TNF|CachectinProtein/peptide typeRecombinantProtein FunctionCytokineSpeciesMouseSourceE. coliConjugationUnconjugatedGene IDP06804SequenceMLRSSSQNSS DKPVAHVVAN HQVEEQLEWL SQRANALLAN GMDLKDNQLV VPADGLYLVY SQVLFKGQGC PDYVLLTHTV SRFAISYQEK VNLLSAVKSP CPKDTPEGAE LKPWYEPIYL GGVFQLEKGD QLSAEVNLPK YLDFAESGQV YFGVIALAmino acid number157Protease-freeNAnimal-freeYNuclease-freeNCarrier-freeYEndotoxin-freeNEndotoxin levelLowEndotoxin contentEndotoxin LAL (EU/µg) ≤0.1Biological activityBioactivity Assay: Cytolysis of mouse L929 cells in the presence of Actinomycin D, ED₅₀ ≤0.1 ng/ml, Bioactivity ≥1.0×10⁷ units/mgPurity≥95%GradeRUOFormulation10 mM sodium phosphate, 50 mM sodium chloride, pH 7.5Molecular weight17.4 kDaReconstitution instructionsSterile water at 0.1 mg/mlStorage conditions–20 °CShipping temperatureRTTested applicationsBioassay - Supplier Number: 200-31AF-1MGSpecifications:
More
Specifications
Cat. No.77660-834Protein/peptide nameTNF-A (AF)Protein synonymsCytotoxin|DIF|Necrosin|Cachexin|TNFSF2|TNF|CachectinProtein/peptide typeRecombinantProtein FunctionCytokineSpeciesMouseSourceE. coliConjugationUnconjugatedGene IDP06804SequenceMLRSSSQNSS DKPVAHVVAN HQVEEQLEWL SQRANALLAN GMDLKDNQLV VPADGLYLVY SQVLFKGQGC PDYVLLTHTV SRFAISYQEK VNLLSAVKSP CPKDTPEGAE LKPWYEPIYL GGVFQLEKGD QLSAEVNLPK YLDFAESGQV YFGVIALAmino acid number157Protease-freeNAnimal-freeYNuclease-freeNCarrier-freeYEndotoxin-freeNEndotoxin levelLowEndotoxin contentEndotoxin LAL (EU/µg) ≤0.1Biological activityBioactivity Assay: Cytolysis of mouse L929 cells in the presence of Actinomycin D, ED₅₀ ≤0.1 ng/ml, Bioactivity ≥1.0×10⁷ units/mgPurity≥95%GradeRUOFormulation10 mM sodium phosphate, 50 mM sodium chloride, pH 7.5Molecular weight17.4 kDaReconstitution instructionsSterile water at 0.1 mg/mlStorage conditions–20 °CShipping temperatureRTTested applicationsBioassay
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



