Order Entry
Canada
ContactUsLinkComponent
Human Recombinant FGF-8 (from E. coli)
Human Recombinant FGF-8 (from E. coli)
New Product
Certificates
Human Recombinant FGF-8 (from E. coli)
New Product

Specifications

  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokine
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    At or below −20 °C
  • Endotoxin content:
    Endotoxin LAL (EU/µg) ≤0.1
  • Biological activity:
    Bioactivity assay: NR6R 3T3 cell proliferation. ED50 ≤150 ng/ml. bioactivity ≥6.7×10³ units/mg
  • Gene ID:
    P55075/P37237
  • Reconstitution instructions:
    Sterile water at 0.1 mg/ml
  • Endotoxin-free:
    N
  • Carrier-free:
    Y
  • Protease-free:
    N
  • Animal-free:
    N
  • Protein synonyms:
    Androgen-induced growth factor|AIGF|FGF-8b isoform|Heparin-binding growth factor 8|HBGF-8
  • Accession no.:
    P55075/P37237
  • Protein/peptide name:
    FGF-8
  • Purity:
    ≥95%
  • Molecular weight:
    22.5 kDa
  • Sequence:
    MQVTVQSSPN FTQHVREQSL VTDQLSRRLI RTYQLYSRTS GKHVQVLANK RINAMAEDGD PFAKLIVETD TFGSRVRVRG AETGLYICMN KKGKLIAKSN GKGKDCVFTE IVLENNYTAL QNAKYEGWYM AFTRKGRPRK GSKTRQHQRE VHFMKRLPRG HHTTEQSLRF EFLNYPPFTR SLRGSQRTWA PEPR
  • Endotoxin level:
    Low
  • Formulation:
    0.1% Trifluoroacetic Acid (TFA)
  • Nuclease-free:
    N
  • Grade:
    RUO
  • Shipping temperature:
    RT
  • Tested applications:
    Bioassay

Specifications

Frequently Bought Together