Orders LinkContactUsLinkComponent
Crile Hemostatic Forceps
Crile Hemostatic Forceps
Crile Hemostatic Forceps
Certificates
Crile Hemostatic Forceps
9 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Crile hemostatic forceps are atraumatic and non-toothed clamps used to clamp blood vessels, grab tissue or vessels.
They are often known as a 'snap.'
Product Family Options
Product Information
  • Material
    Length
    Tip style
    Availability
    Price
  • Stainless steel
    14.0 cm (5.5")
    Straight
    Log in for availability
    Log in for availability
    Each
    Each - $92.46
    $92.46
    Each
       $92.46
    Supplier Number: 501242
  • Stainless steel
    14.0 cm (5.5")
    Curved
    Log in for availability
    Log in for availability
    Each
    Each - $92.46
    $92.46
    Each
       $92.46
    Supplier Number: 501730
  • German steel
    14.0 cm (5.5")
    Curved
    Log in for availability
    Log in for availability
    Each
    Each - $83.74
    $83.74
    Each
       $83.74
    Supplier Number: 501730-G
  • Stainless steel
    14.0 cm (5.5")
    Straight/smooth
    Log in for availability
    Log in for availability
    Each
    Each - $60.85
    $60.85
    Each
       $60.85
    Supplier Number: 503227
  • Stainless steel
    14.0 cm (5.5")
    Curved/smooth
    Log in for availability
    Log in for availability
    Each
    Each - $60.85
    $60.85
    Each
       $60.85
    Supplier Number: 503228
  • Stainless steel
    15.9 cm (6.25")
    Straight
    Log in for availability
    Log in for availability
    Each
    Each - $65.71
    $65.71
    Each
       $65.71
    Supplier Number: 501712
  • Stainless steel
    15.9 cm (6.25")
    Curved
    Log in for availability
    Log in for availability
    Each
    Each - $65.71
    $65.71
    Each
       $65.71
    Supplier Number: 501713
  • German steel
    15.9 cm (6.25")
    Curved
    Log in for availability
    Log in for availability
    Each
    Each - $83.74
    $83.74
    Each
       $83.74
    Supplier Number: 501713-G
  • Stainless steel
    15.9 cm (6.25")
    Straight/smooth
    Log in for availability
    Log in for availability
    Each
    Each - $60.85
    $60.85
    Each
       $60.85
    Supplier Number: 503230
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)