Orders LinkContactUsLinkComponent
Microdissecting Forceps
Microdissecting Forceps
Microdissecting Forceps
Certificates
Microdissecting Forceps
4 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Microdissecting forceps have fine tips for precision work in microsurgery, microdissection, and applications requiring fine manipulation of tissue or small objects under a microscope.
  • Angled, angled fine, straight fine tips
  • Surgical grade stainless steel
The tips are only 0.8 mm wide, and you can get straight or angled tips. These are hand forceps with a spring action letting you pinch the blades together to control the amount of pressure used. The material is suitable for autoclaving or chemical sterilization. Clean surgical forceps after use to prevent staining and increase their useable life.
Product Family Options
Product Information
  • Length
    Tip style
    Tip width
    Tip configuration
    Availability
    Price
  • 77663-604
    Microdissecting forceps
    Microdissecting forceps
    10.2 cm (4")
    Serrated
    0.8 mm
    Angled
    Log in for availability
    Log in for availability
    Each
    Each - $92.46
    $92.46
    Each
       $92.46
    Supplier Number: 504479
  • 77663-608
    Microdissecting forceps
    Microdissecting forceps
    10.2 cm (4")
    Smooth, sharp
    0.8 mm
    Angled fine
    Log in for availability
    Log in for availability
    Each
    Each - $133.83
    $133.83
    Each
       $133.83
    Supplier Number: 504481
  • 77663-610
    Microdissecting forceps
    Microdissecting forceps
    10.2 cm (4")
    Sharp
    0.8 mm
    Angled fine
    Log in for availability
    Log in for availability
    Each
    Each - $133.83
    $133.83
    Each
       $133.83
    Supplier Number: 504482
  • 77663-612
    Microdissecting forceps
    Microdissecting forceps
    10.2 cm (4")
    Sharp
    0.8 mm
    Straight fine
    Log in for availability
    Log in for availability
    Each
    Each - $131.39
    $131.39
    Each
       $131.39
    Supplier Number: 504483
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)