Orders LinkContactUsLinkComponent
Foerster Forceps
Foerster Forceps
Foerster Forceps
Certificates
Foerster Forceps
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Foerster Forceps, also known as Foerster sponge forceps or Foerster-Ballenger forceps, are a surgical instrument commonly used in medical procedures like gynecology and obstetrics. These forceps are named after the German surgeon Heinrich Foerster. The primary purpose of Foerster forceps is to grasp and hold surgical sponges or other materials.
  • Surgical grade stainless steel
The serrated jaws help prevent slippage and ensure a firm grip. The easily recognizable, hexagon-shaped handles aids in distinguishing and handling this instrument, differentiating it from other surgical forceps. This handle also provides a secure grip, especially when you are wearing gloves or your hands are slippery. The hexagonal handle prevents the forceps from rolling when you set them down on a flat surface.
Choose either a curved or a straight, serrated tip. Material suitable for autoclaving or chemical sterilization. Always clean surgical instruments promptly after use to prevent staining and ensure a longer useable life.
Product Family Options
Product Information
  • Material
    Length
    Tip style
    Tip width
    Curvature
    Availability
    Price
  • Stainless steel
    10.2 cm (4")
    Serrated
    0.5 mm
    Curved
    Log in for availability
    Log in for availability
    Each
    Each - $240.90
    $240.90
    Each
       $240.90
    Supplier Number: 504477
  • Stainless steel
    10.2 cm (4")
    Serrated
    0.5 mm
    Straight
    Log in for availability
    Log in for availability
    Each
    Each - $233.60
    $233.60
    Each
       $233.60
    Supplier Number: 504476
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)