If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Description:Demagnetize/magnetize surgical instruments, 2×2×1"
- Cat. no.:77662-944
Specifications
About this item
Instantly magnetize or demagnetize small surgical instruments and small steel parts that are over 1.3" in length and 0.125" in diameter.
- No batteries or electricity required
- Box is high quality molded ABS plastic, impact resistant and cadmium free
- The innovative step design lets you control the level of magnetism
To Magnetise a tool: Insert tool into the "+" opening and slide back and forth. Continue this procedure until the desired level of magnetism is reached.
To Demagnetise a tool: Insert tool at the lowest "−" opening. Slide back and forth. Continue the procedure, moving up to the next step until the tool is completely demagnetised.