AccuVac® Test Kits, Photometric
104088-462
: Hach
:
- Chemical method:DPD
- Description:Chlorine (free) DPD
- Cat. no.:104088-462
- Range:0.02 - 2.00 mg/L
- Type:AV
AV - AccuVac® - Sealed glass cuvettes containing a portioned amount of reagent. Simply immerse the tip of the accuvac in the sample, break it and the vacuum draws the sample into the cuvette.