ContactUsLinkComponent
Comparator Kits for Water Testing
Comparator Kits for Water Testing
Catalog # 104085-380
Supplier:  Hach
CAS Number:  
undefined
Comparator Kits for Water Testing
Catalog # 104085-380
Supplier:  Hach
Supplier Number:  1419301
CAS Number:  
Specifications
  • Description:
    Molybdate Test Kit, Model MO-2
  • Cat. no.:
    104085-380
  • Range:
    0 - 0.8 / 0 - 40 mg/L
  • No. of Tests:
    100
Specifications
About this item
Qualitative results using traditional color comparator disc methods.
  • Portable - ideal for field testing
  • Simple fast method
  • Kits contain all required reagents and apparatus in a rugged carrying case
Please note some of the reagents supplied in the kits are hazardous, and appropriate precautions should be taken for use and disposal. Please contact Avantor for details.