Reagent Sets for Measurement of Phosphates and ortho-Phosphates - Test 'N Tube Method
104088-456
: Hach
:
- Chemical method:PhosVer3 - Molybdenum blue
- Description:Phosphate (ortho), low range Test 'N Tube reagent set, with PhosVer® 3 reagent powder pillows and 'reactive' phosphorus Test ‘N Tube vials
- Cat. no.:104088-456
- Range:0.06 - 5 mg/L
- No. of Tests:50
- For Unit:DR900
Suitable for measurement with selected Hach photometers which are suitable for 16 mm tubes.
- Convenient testing in tube, with minimal handling of chemicals
- Digestion (where required) is performed in the same tube as the test
: The reagent sets may contain materials which are hazardous. Please read the usage instructions and MSDS for details of handling and disposal.