Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Canada
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Environmental Test Kits
Powder Pillows, Iron
Powder Pillows, Iron
104087-034
:
Hach
:
Powder Pillows, Iron
104087-034
:
Hach
:
92746
:
Chemical method:
1,10-Phenanthroline
Description:
Iron (FerroVer®) reagent
Cat. no.:
104087-034
Range:
0 - 10 mg/L
Type:
PP
Sample Size:
5 ml