Silica Reagents for 5000 Silica Analyzer
104088-460
: Hach
:
- Description:Molybdate 3 reagent
- Cat. no.:104088-460
- For Unit:S5000 Silica
One unit each of the reagents is sufficient for 30 days of operation. The standard solution is sufficient for ten calibrations.