ContactUsLinkComponent
IntelliCAL™ MTC101/MTC301 ORP Combination Electrodes
IntelliCAL™ MTC101/MTC301 ORP Combination Electrodes
Catalog # 104087-744
Supplier:  Hach
undefined
IntelliCAL™ MTC101/MTC301 ORP Combination Electrodes
Catalog # 104087-744
Supplier:  Hach
Supplier Number:  MTC30101
Specifications
  • Cat. no.:
    104087-744
  • For Use With:
    Wastewater, drinking water, general applications
  • Junction:
    Ceramic pin
  • mV Range:
    ±1200
  • Connector:
    1 m cable
  • Electrolyte:
    3.0 M KCl (refillable)
Specifications
About this item
Digital combination ORP/Redox probes with built-in temperature sensor. Ideal for measuring absolute mV values in wastewater, drinking water and general applications.
  • Compatible with digital Hach meters only (e.g. HQd meters)
  • Traceability in measurement history: Stored time and date stamp for each measurement, operator and sample ID, calibration history, parameter, and probe serial number
  • User alerted when re-calibration is needed
  • Plug & Play: Can be moved between meters without the need to re-calibrate or re-enter measurement settings
  • Rugged digital probes can reliably test in the harshest environmental conditions
Available with a non refillable gel electrolyte (MTC101) or refillable 3M KCl electrolyte (MTC301).
The standard versions are available with a 1 or 3 meter cable and are intended for laboratory use. The rugged versions are available with a 5, 10 or 15 meter cable and are intended for field use.