ContactUsLinkComponent
AccuTec Pro® Single Edge Blades - Round Corner, AccuTec Blades
AccuTec Pro® Single Edge Blades - Round Corner, AccuTec Blades
Catalog # 77676-540
Supplier:  Accutec Blades
New Product
undefined
AccuTec Pro® Single Edge Blades - Round Corner, AccuTec Blades
Catalog # 77676-540
Supplier:  Accutec Blades
Supplier Number:  APBL-7041-0000
New Product
Specifications
  • Cat. no.:
    77676-540
  • Backing Material:
    Aluminum
Specifications
About this item
AccuTecPro Single Edge blades feature precision ground edges for sharpness and durability. Blades are unwrapped for high volume use.
  • Round corner design for safety
  • 2-facet edge for increased durability
  • Heavy duty .012" carbon steel
  • Bulk, unwrapped blades are ideal for high volume use
  • Each box contains 225 blades
  • Made in USA
Caution: Blades are extremely sharp. Wear eye protection. Handle and use with care. Bending blades may break them. Keep out of children’s reach. Dispose of safely.