Order Entry
Canada
ContactUsLinkComponent
Lid Strips, Optimized for qPCR
Lid Strips, Optimized for qPCR
Catalog # 77588-352
Supplier:  SARSTEDT INC
New Product
undefined
Lid Strips, Optimized for qPCR
Catalog # 77588-352
Supplier:  SARSTEDT INC
Supplier Number:  65.989
New Product

Specifications

  • Color:
    Transparent
  • Description:
    Lid strips, optimised for qPCR, Biosphere® plus
  • Cat. no.:
    77588-352

Specifications

About this item

SARSTEDT PCR consumables are compatible with standard thermocyclers. Uniform and extremely thin well walls facilitate consistently homogeneous and rapid heat transfer. During production, each individual well is tested to ensure it is 100% leak-proof.

  • Polypropylene material
  • Highly transparent for efficient light transmission
  • Two purity standards offer tailored product quality
  • For use with PCR strips and PCR plates

SARSTEDT PCR tubes are 'PCR Performance Tested' certified and guaranteed to be DNA-free, free from DNase/RNase, and free from PCR inhibitors. For the absolutely reliable absence of DNA and other biomolecules, Biosphere® plus certified products are subjected to a validated decontamination procedure. Biosphere® plus products are also free from DNA, DNase/RNase, PCR inhibitors, ATP, non-pyrogenic/endotoxin-free and are supplied sterile.