Pneumatic Quick-Disconnect Lockout, Brady Worldwide®
Supplier: Brady Worldwide
Certificates
About this item
Isolates pneumatic energy without the expense and inconvenience of installing in-line lockout valves
Device is applied to the male fitting, isolating equipment from all compressed air sources. Accommodates the majority of 6mm (1/4"), 10mm (3/8"), and 13mm (1/2") male fittings. Hole in center of device allows for permanent storage on an air hose and a loop on the side can be used to hang the hose and lockout device. Lockout accepts padlocks with 6mm (1/4") or 7mm (9/32") shackles.
Customers Who Bought This Also Bought