Orders LinkContactUsLinkComponent
Prinzing® Plug Lockout, Brady®
Prinzing® Plug Lockout, Brady®
Plug Lockout
Catalog # CA56606-578
Supplier:   Brady Worldwide
Supplier #:  PLO23
Prinzing® Plug Lockout, Brady®
Catalog # CA56606-578
Supplier:  Brady Worldwide
Prinzing® Plug Lockout, Brady®
Prinzing® Plug Lockout, Brady®
Plug Lockout
Price
$26.55
Log in for availability
Each
Product Details & Documents
Safely prevents machinery from being plugged in during cleaning and maintenance by preventing use of plug in an electrical outlet.
Locks out any electrical plug up to 7.6Wx14Lcm (3x51/2") with a maximum cord diameter of 3.8cm (11/2"). Made of bright yellow thermoplastic, the plug lockout gives a visual warning when in use. Two hinged over-plates allow flexibility in accommodating various cord diameters.
Specifications
  • Description:
    Plug Lockout