Orders LinkContactUsLinkComponent
Operating Scissors, 18 cm
Operating Scissors, 18 cm
Operating Scissors, 18 cm
Certificates
Operating Scissors, 18 cm
5 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Ring-handled Operating Scissors are a pair of surgical scissors commonly used in general surgery for cutting and dissecting tissues.
  • Autoclavable
  • Straight or curved
  • Sharp/sharp, Sharp/blunt, or Blunt/blunt tips
  • Stainless steel or premium German steel
The 18 cm length is a versatile size that can be used for a variety of procedures, including biological and medical research. Operating scissors are for general use in dissection, preparation of tissue samples, and small animal surgeries. These surgical scissors are available with a variety of tips, including two sharp pointed blades, one sharp and one blunt blade, or two blunt blades.
They are available in both surgical grade stainless steel and our premium German steel, making them more durable and resistant to corrosion. Operating Scissors are designed to withstand the high heat and chemical exposure of the sterilization process.
Product Family Options
Product Information
  • Length
    Material
    Curvature
    Availability
    Price
  • 77661-962
    Operating scissors, sharp/sharp
    Operating scissors, sharp/sharp
    18 cm (7")
    Stainless steel
    Straight
    Log in for availability
    Log in for availability
    Each
    Each - $92.46
    $92.46
    Each
       $92.46
    Supplier Number: 15924
  • 77661-964
    Operating scissors, sharp/sharp, German steel
    Operating scissors, sharp/sharp, German steel
    18 cm (7")
    Premium German steel
    Straight
    Log in for availability
    Log in for availability
    Each
    Each - $104.30
    $104.30
    Each
       $104.30
    Supplier Number: 15924-G
  • 77662-330
    Operating scissors, sharp/blunt
    Operating scissors, sharp/blunt
    18 cm (7")
    Stainless steel
    Straight
    Log in for availability
    Log in for availability
    Each
    Each - $92.46
    $92.46
    Each
       $92.46
    Supplier Number: 500378
  • 77662-332
    Operating scissors, sharp/blunt
    Operating scissors, sharp/blunt
    18 cm (7")
    Premium German steel
    Straight
    Log in for availability
    Log in for availability
    Each
    Each - $97.94
    $97.94
    Each
       $97.94
    Supplier Number: 500378-G
  • 77662-334
    Operating scissors, blunt/blunt
    Operating scissors, blunt/blunt
    18 cm (7")
    Premium German steel
    Straight
    Log in for availability
    Log in for availability
    Each
    Each - $104.30
    $104.30
    Each
       $104.30
    Supplier Number: 500379-G
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)