Orders LinkContactUsLinkComponent
Operating Scissors
Operating Scissors
Operating Scissors
Certificates
Operating Scissors
31 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Ring-handled operating scissors are operating scissors are available with straight or curved blades commonly used in general surgery for cutting and dissecting tissues.
  • Sharp/sharp, Sharp/blunt, or Blunt/blunt tips
  • Stainless steel or premium German steel
  • Autoclavable
That can be used for a variety of procedures, including biological and medical research. Our operating scissors are for general use in dissection, preparation of tissue samples, and small animal surgeries. These surgical scissors are available with a variety of tips, including two sharp pointed blades, one sharp and one blunt blade, or two blunt blades.
They are available in both surgical grade stainless steel and our premium German steel, making them more durable and resistant to corrosion. Operating scissors are designed to withstand the high heat and chemical exposure of the sterilization process.
Product Family Options
Straight
Product Information
  • Length
    Material
    Availability
    Price
  • 77661-820
    Operating scissors
    Operating scissors
    14 cm (5.5")
    Stainless steel
    Log in for availability
    Log in for availability
    Each
    Each - $92.46
    $92.46
    Each
       $92.46
    Supplier Number: 14192
    Specifications:
    More
    Specifications
    Cat. No.
    77661-820
  • 77661-822
    Operating scissors
    Operating scissors
    14 cm (5.5")
    Premium German steel
    Log in for availability
    Log in for availability
    Each
    Each - $82.16
    $82.16
    Each
       $82.16
    Supplier Number: 14192-G
    Specifications:
    More
    Specifications
    Cat. No.
    77661-822
  • 77662-396
    Operating scissors
    Operating scissors
    14 cm (5.5")
    Stainless steel
    Log in for availability
    Log in for availability
    Each
    Each - $92.46
    $92.46
    Each
       $92.46
    Supplier Number: 501218
    Specifications:
    More
    Specifications
    Cat. No.
    77662-396
  • 77662-398
    Operating scissors
    Operating scissors
    14 cm (5.5")
    Premium German steel
    Log in for availability
    Log in for availability
    Each
    Each - $82.16
    $82.16
    Each
       $82.16
    Supplier Number: 501218-G
    Specifications:
    More
    Specifications
    Cat. No.
    77662-398
  • 77662-400
    Operating scissors
    Operating scissors
    14 cm (5.5")
    Stainless steel
    Log in for availability
    Log in for availability
    Each
    Each - $92.46
    $92.46
    Each
       $92.46
    Supplier Number: 501219
    Specifications:
    More
    Specifications
    Cat. No.
    77662-400
  • 77662-402
    Operating scissors
    Operating scissors
    14 cm (5.5")
    Premium German steel
    Log in for availability
    Log in for availability
    Each
    Each - $71.08
    $71.08
    Each
       $71.08
    Supplier Number: 501219-G
    Specifications:
    More
    Specifications
    Cat. No.
    77662-402
  • 77662-416
    Operating scissors
    Operating scissors
    16 cm (6.25")
    Stainless steel
    Log in for availability
    Log in for availability
    Each
    Each - $92.46
    $92.46
    Each
       $92.46
    Supplier Number: 501223
    Specifications:
    More
    Specifications
    Cat. No.
    77662-416
  • 77662-418
    Operating scissors
    Operating scissors
    16 cm (6.25")
    Premium German steel
    Log in for availability
    Log in for availability
    Each
    Each - $79.00
    $79.00
    Each
       $79.00
    Supplier Number: 501223-G
    Specifications:
    More
    Specifications
    Cat. No.
    77662-418
  • 77662-420
    Operating scissors
    Operating scissors
    16 cm (6.25")
    Stainless steel
    Log in for availability
    Log in for availability
    Each
    Each - $92.46
    $92.46
    Each
       $92.46
    Supplier Number: 501224
    Specifications:
    More
    Specifications
    Cat. No.
    77662-420
  • 77662-422
    Operating scissors
    Operating scissors
    16 cm (6.25")
    Premium German steel
    Log in for availability
    Log in for availability
    Each
    Each - $79.00
    $79.00
    Each
       $79.00
    Supplier Number: 501224-G
    Specifications:
    More
    Specifications
    Cat. No.
    77662-422
  • 77662-424
    Operating scissors
    Operating scissors
    16 cm (6.25")
    Stainless steel
    Log in for availability
    Log in for availability
    Each
    Each - $92.46
    $92.46
    Each
       $92.46
    Supplier Number: 501225
    Specifications:
    More
    Specifications
    Cat. No.
    77662-424
  • 77662-426
    Operating scissors
    Operating scissors
    16 cm (6.25")
    Premium German steel
    Log in for availability
    Log in for availability
    Each
    Each - $79.00
    $79.00
    Each
       $79.00
    Supplier Number: 501225-G
    Specifications:
    More
    Specifications
    Cat. No.
    77662-426
  • 77662-764
    Operating scissors
    Operating scissors
    11.5 cm (4.5")
    Stainless steel
    Log in for availability
    Log in for availability
    Each
    Each - $90.03
    $90.03
    Each
       $90.03
    Supplier Number: 501743
    Specifications:
    More
    Specifications
    Cat. No.
    77662-764
  • 77662-766
    Operating scissors
    Operating scissors
    11.5 cm (4.5")
    Premium German steel
    Log in for availability
    Log in for availability
    Each
    Each - $82.16
    $82.16
    Each
       $82.16
    Supplier Number: 501743-G
    Specifications:
    More
    Specifications
    Cat. No.
    77662-766
  • 77662-800
    Operating scissors
    Operating scissors
    11.5 cm (4.5")
    Stainless steel
    Log in for availability
    Log in for availability
    Each
    Each - $90.03
    $90.03
    Each
       $90.03
    Supplier Number: 501753
    Specifications:
    More
    Specifications
    Cat. No.
    77662-800
  • 77662-802
    Operating scissors
    Operating scissors
    11.5 cm (4.5")
    Premium German steel
    Log in for availability
    Log in for availability
    Each
    Each - $82.16
    $82.16
    Each
       $82.16
    Supplier Number: 501753-G
    Specifications:
    More
    Specifications
    Cat. No.
    77662-802
  • 77662-804
    Operating scissors
    Operating scissors
    11.5 cm (4.5")
    Stainless steel
    Log in for availability
    Log in for availability
    Each
    Each - $90.03
    $90.03
    Each
       $90.03
    Supplier Number: 501754
    Specifications:
    More
    Specifications
    Cat. No.
    77662-804
  • 77662-806
    Operating scissors
    Operating scissors
    11.5 cm (4.5")
    Premium German steel
    Discontinued
    Supplier Number: 501754-G
    Specifications:
    More
    Specifications
    Cat. No.
    77662-806
Curved
Product Information
  • Length
    Material
    Availability
    Price
  • 77662-404
    Operating scissors
    Operating scissors
    14 cm (5.5")
    Stainless steel
    Log in for availability
    Log in for availability
    Each
    Each - $92.46
    $92.46
    Each
       $92.46
    Supplier Number: 501220
    Specifications:
    More
    Specifications
    Cat. No.
    77662-404
  • 77662-406
    Operating scissors
    Operating scissors
    14 cm (5.5")
    Premium German steel
    Log in for availability
    Log in for availability
    Each
    Each - $86.90
    $86.90
    Each
       $86.90
    Supplier Number: 501220-G
    Specifications:
    More
    Specifications
    Cat. No.
    77662-406
  • 77662-408
    Operating scissors
    Operating scissors
    14 cm (5.5")
    Stainless steel
    Log in for availability
    Log in for availability
    Each
    Each - $92.46
    $92.46
    Each
       $92.46
    Supplier Number: 501221
    Specifications:
    More
    Specifications
    Cat. No.
    77662-408
  • 77662-410
    Operating scissors
    Operating scissors
    14 cm (5.5")
    Premium German steel
    Log in for availability
    Log in for availability
    Each
    Each - $82.16
    $82.16
    Each
       $82.16
    Supplier Number: 501221-G
    Specifications:
    More
    Specifications
    Cat. No.
    77662-410
  • 77662-412
    Operating scissors
    Operating scissors
    14 cm (5.5")
    Stainless steel
    Log in for availability
    Log in for availability
    Each
    Each - $92.46
    $92.46
    Each
       $92.46
    Supplier Number: 501222
    Specifications:
    More
    Specifications
    Cat. No.
    77662-412
  • 77662-414
    Operating scissors
    Operating scissors
    14 cm (5.5")
    Premium German steel
    Log in for availability
    Log in for availability
    Each
    Each - $79.00
    $79.00
    Each
       $79.00
    Supplier Number: 501222-G
    Specifications:
    More
    Specifications
    Cat. No.
    77662-414
  • 77662-428
    Operating scissors
    Operating scissors
    16 cm (6.25")
    Stainless steel
    Log in for availability
    Log in for availability
    Each
    Each - $92.46
    $92.46
    Each
       $92.46
    Supplier Number: 501226
    Specifications:
    More
    Specifications
    Cat. No.
    77662-428
  • 77662-430
    Operating scissors
    Operating scissors
    16 cm (6.25")
    Premium German steel
    Log in for availability
    Log in for availability
    Each
    Each - $79.00
    $79.00
    Each
       $79.00
    Supplier Number: 501226-G
    Specifications:
    More
    Specifications
    Cat. No.
    77662-430
  • 77662-432
    Operating scissors
    Operating scissors
    16 cm (6.25")
    Premium German steel
    Discontinued
    Supplier Number: 501227-G
    Specifications:
    More
    Specifications
    Cat. No.
    77662-432
  • 77662-434
    Operating scissors
    Operating scissors
    16 cm (6.25")
    Stainless steel
    Log in for availability
    Log in for availability
    Each
    Each - $90.03
    $90.03
    Each
       $90.03
    Supplier Number: 501228
    Specifications:
    More
    Specifications
    Cat. No.
    77662-434
  • 77662-436
    Operating scissors
    Operating scissors
    16 cm (6.25")
    Premium German steel
    Log in for availability
    Log in for availability
    Each
    Each - $66.36
    $66.36
    Each
       $66.36
    Supplier Number: 501228-G
    Specifications:
    More
    Specifications
    Cat. No.
    77662-436
  • 77662-438
    Operating scissors
    Operating scissors
    18 cm (7")
    Stainless steel
    Log in for availability
    Log in for availability
    Each
    Each - $65.71
    $65.71
    Each
       $65.71
    Supplier Number: 501229
    Specifications:
    More
    Specifications
    Cat. No.
    77662-438
  • 77662-440
    Operating scissors
    Operating scissors
    18 cm (7")
    Premium German steel
    Log in for availability
    Log in for availability
    Each
    Each - $88.48
    $88.48
    Each
       $88.48
    Supplier Number: 501229-G
    Specifications:
    More
    Specifications
    Cat. No.
    77662-440
  • 77662-442
    Operating scissors
    Operating scissors
    18 cm (7")
    Stainless steel
    Log in for availability
    Log in for availability
    Each
    Each - $94.91
    $94.91
    Each
       $94.91
    Supplier Number: 501230
    Specifications:
    More
    Specifications
    Cat. No.
    77662-442
  • 77662-444
    Operating scissors
    Operating scissors
    18 cm (7")
    Stainless steel
    Log in for availability
    Log in for availability
    Each
    Each - $92.46
    $92.46
    Each
       $92.46
    Supplier Number: 501231
    Specifications:
    More
    Specifications
    Cat. No.
    77662-444
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)