Orders LinkContactUsLinkComponent
Microelectrode Holders
Microelectrode Holders
Microelectrode Holders
WPI™s microelectrode holder-half-cells couple fluid-filled glass micropipettes to high input impedance amplifiers. A Ag/AgCl pellet (or a silver wire) molded into the holder body provides stable potential. Electrical connection is made via male 2 mm pins or female 2 mm sockets.
  • Male connector
  • Pellet half-cell
  • No pressure port, no cap
The pipette may be mounted axially or at right angles to the holder. Pipettes are held with screw-caps or rubber gaskets (without caps). Filling WPI microelectrode holders with electrolytes containing chloride results in stable electrode potential. Suitable electrolytes include KCl, NaCl and CaCl₂. Holders are supplied for standard WPI single capillary tubing of 1.0, 1.2, 1.5 and 2.0 mm outside diameters.
Product Family Options
  • For use with
  • 1.0 mm standard WPI single capillary tubing
    Each
    Each - $142.20
    $142.20
    Each
       $142.20
     : MEH1R10
  • 1.2 mm standard WPI single capillary tubing
    Each
    Each - $142.20
    $142.20
    Each
       $142.20
     : MEH1R12
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.