ContactUsLinkComponent
Stablcal® Turbidity Standards
  104088-274
 :  Hach
 :  
undefined
Stablcal® Turbidity Standards
  104088-274
 :  Hach
 :  2660649
 :  
  • Turbidity:
    1000 NTU
  • Description:
    Stablcal® stabilized formazine standard
  • Size:
    500 ml
  • Cat. no.:
    104088-274
Stablcal® turbidity standards for all turbidimeters. USEPA accepted for calibration purposes. Stablcal® stabilized formazine turbidity standards are true formazine dilutions developed for use in any turbidimeter.
  • Precise, pre-mixed formazine solutions
  • Based on formazine primary standards
  • Enhanced stability saves time and labour
  • Confidence in results