Test Kits, Digital Titration Method
Catalog # 104088-218
Supplier: Hach
CAS Number:
Specifications
- Description:Hardness (Calcium) reagent set, low range
- Cat. no.:104088-218
- Range:10 - 160 mg/L
Specifications
About this item
Digital titration provides a more precise and accurate method than the drop test method.
- Reagents are pre-prepared
- Exact comparison of color
- Rugged carrying case for analysis in the field
Caution: Please note some of the reagents supplied in the kits are hazardous, and appropriate precautions should be taken for use and disposal. Please contact Avantor for details.