These standard glass capillaries are made with quality, borosilicate glass and used for applications such as: Microinjection, electrophysiology, patch clamp, and fluid handling.
- Quality borosilicate glass capillaries
- Large variety available
- Some varieties are fire polished
- OD/ID tolerance: ±0.1 mm
- Length tolerance: ±1 mm
- Recommended to use with glass handling forceps
Fire-Polished glass capillaries are easier to insert into microelectrode holders without damaging the gasket. More importantly, fire-polished glass won’t scratch the chloridized wire used in a recording electrode. Fire-polishing does not affect the glass’s mechanical or electrical properties.
Single barrel standard wall thickness capillaries are offered either with or without inner filaments for quick filling in a variety of lengths and diameters. Thin Wall single barrel capillaries are offered both with or without inner filaments.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.