SHOWA 719B Nitrile, Chemical Resistant Gloves
Supplier: Showa
New Product
Certificates
About this item
Protecting against a broad range of hazards, the SHOWA 719B chemical-resistant gloves features an extended gauntlet for forearm protection.
- Chemical-resistant
- Water-resistant
- Abrasion-resistant
- Forearm protection
- Oil-resistant
The 0.28 mm thick unsupported nitrile provides resistance against chemicals, solvents, oils, and animal fats. The ergonomic design ensures comfort during long periods of wear.
Featuring a diamond grip surface makes wet work easier and safer.
Specifications
- Catalog No:
- 77623-468
- 77623-474
- 77623-472
- 77623-470
- 77623-466
- Disposable / reusable:
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Liner material:
- Unlined
- Unlined
- Unlined
- Unlined
- Unlined
- Material:
- Nitrile
- Nitrile
- Nitrile
- Nitrile
- Nitrile