Orders LinkContactUsLinkComponent
Dressing Forceps
Dressing Forceps
Certificates
Dressing Forceps
9 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Dressing forceps, also known as thumb forceps or pickups, are surgical instruments characterised by their long, thin handles and serrated tips. They are typically used when dressing wounds where they hold gauze and other dressings.
  • Dressing forceps with straight blades
  • Serrated jaws
  • Straight tips
  • Surgical grade stainless steel or premium German steel
They may also be used during wound debridement to remove infected or necrotic tissue or debris from the wound. They may be used for suturing when needle holders are not accessible. Dressing forceps are designed to grip delicate tissue without causing damage, and they are often used in situations where precision and control are important. These dressing forceps have straight blades and rounded (blunt) tips.
These dressing forceps are manufactured with surgical grade stainless steel or premium German steel, making them suitable for sterilization by autoclave or chemical sterilants. Be sure to clean your instruments properly after every use to remove proteinous and biological debris. Properly cleaning helps to improve the useable lifespan of your surgical instruments.
Product Family Options
Product Information
  • Material
    Length
    Availability
    Price
  • 77662-392
    Dressing forceps, straight
    Dressing forceps, straight
    Stainless steel
    12.5 cm
    Log in for availability
    Log in for availability
    Each
    Each - $94.91
    $94.91
    Each
       $94.91
    Supplier Number: 501217
  • 77662-306
    Dressing forceps, straight
    Dressing forceps, straight
    Stainless steel
    15.5 cm
    Log in for availability
    Log in for availability
    Each
    Each - $87.60
    $87.60
    Each
       $87.60
    Supplier Number: 500363
  • 77662-310
    Dressing forceps, straight
    Dressing forceps, straight
    Stainless steel
    25.5 cm
    Log in for availability
    Log in for availability
    Each
    Each - $60.85
    $60.85
    Each
       $60.85
    Supplier Number: 500364
  • 77662-314
    Dressing forceps, straight
    Dressing forceps, straight
    Stainless steel
    30.5 cm
    Log in for availability
    Log in for availability
    Each
    Each - $87.65
    $87.65
    Each
       $87.65
    Supplier Number: 500365
  • 77662-394
    Supercut scissor, straight
    Supercut scissor, straight
    German stainless steel
    12.5 cm
    Log in for availability
    Log in for availability
    Each
    Each - $83.74
    $83.74
    Each
       $83.74
    Supplier Number: 501217-G
  • 77662-308
    Supercut scissor, straight
    Supercut scissor, straight
    German stainless steel
    15.5 cm
    Log in for availability
    Log in for availability
    Each
    Each - $82.16
    $82.16
    Each
       $82.16
    Supplier Number: 500363-G
  • 77662-312
    Supercut scissor, straight
    Supercut scissor, straight
    German stainless steel
    25.5 cm
    Log in for availability
    Log in for availability
    Each
    Each - $83.74
    $83.74
    Each
       $83.74
    Supplier Number: 500364-G
  • 77662-316
    Supercut scissor, straight
    Supercut scissor, straight
    German stainless steel
    30.5 cm
    Log in for availability
    Log in for availability
    Each
    Each - $86.90
    $86.90
    Each
       $86.90
    Supplier Number: 500365-G
  • 77662-736
    Dressing forceps, angled tip
    Dressing forceps, angled tip
    German stainless steel
    15 cm
    Log in for availability
    Log in for availability
    Each
    Each - $113.78
    $113.78
    Each
       $113.78
    Supplier Number: 501727-G
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)