If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Species:HumanMouseNon-human primateRat
- Description:Trypsin in Citrate saline (0.25%)
- Size:100 ml
- Cat. no.:MSPP-07400
Specifications
About this item
0.25% porcine trypsin in citrate saline solution for the detachment of adherent cells.
- Detach adherent cells for easy cell re-suspension and collection
Use Trypsin in Citrate saline (0.25%) for enzymatic cell dissociation. This proteolytic enzyme solution breaks down proteins to detach adherent cells in cell cultures and allows easy resuspension during cell harvesting.